The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000908.3 | NP_000899.1 | 2 - 3 | 1336 | 78 |
NM_001204375.1 | NP_001191304.1 | 2 - 3 | 1336 | 78 | |
NM_001204376.1 | NP_001191305.1 | 2 - 3 | 269 | 78 | |
XM_005248309.1 | XP_005248366.1 | 2 - 3 | 728 | 78 | |
XM_011514047.1 | XP_011512349.1 | 2 - 3 | 233 | 78 | |
XM_011514048.2 | XP_011512350.1 | 2 - 3 | 216 | 78 | |
XM_011514049.2 | XP_011512351.1 | 2 - 3 | 296 | 78 | |
GenBank mRNA | AF025998.1 | - | 2 - 3 | 891 | 78 |
AK290870.1 | - | 2 - 3 | 1341 | 78 | |
AK300094.1 | - | 2 - 3 | 252 | 78 | |
AK308301.1 | - | 2 - 3 | 252 | 78 | |
BC131540.1 | - | 2 - 3 | 1044 | 78 | |
M59305.1 | - | 2 - 3 | 1253 | 78 | |
X52282.1 | - | 2 - 3 | 945 | 78 |