The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000908.3 | NP_000899.1 | 7 - 8 | 1954 | 65 |
NM_001204375.1 | NP_001191304.1 | 7 - 8 | 1957 | 65 | |
NM_001204376.1 | NP_001191305.1 | 7 - 8 | 887 | 65 | |
XM_005248309.1 | XP_005248366.1 | 7 - 8 | 1349 | 65 | |
XM_011514047.1 | XP_011512349.1 | 7 - 8 | 854 | 65 | |
XM_011514048.2 | XP_011512350.1 | 7 - 8 | 837 | 65 | |
XM_011514049.2 | XP_011512351.1 | 7 - 8 | 917 | 65 | |
XM_017009492.1 | XP_016864981.1 | 6 - 7 | 1844 | 65 | |
GenBank mRNA | AF025998.1 | - | 7 - 8 | 1509 | 65 |
AK290870.1 | - | 7 - 8 | 1962 | 65 | |
AK300094.1 | - | 7 - 8 | 870 | 65 | |
BC131540.1 | - | 7 - 8 | 1665 | 65 | |
M59305.1 | - | 7 - 8 | 1874 | 65 | |
X52282.1 | - | 7 - 8 | 1563 | 65 |