The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308028.1 | NP_001294957.1 | 16 - 17 | 2198 | 60 |
NM_001308031.1 | NP_001294960.1 | 9 - 10 | 1148 | 60 | |
NM_005246.3 | NP_005237.2 | 18 - 19 | 2584 | 60 | |
XM_011543266.1 | XP_011541568.1 | 19 - 20 | 2482 | 60 | |
XM_011543267.1 | XP_011541569.1 | 19 - 20 | 4366 | 60 | |
XM_011543269.1 | XP_011541571.1 | 18 - 19 | 4220 | 60 | |
XM_011543270.2 | XP_011541572.1 | 18 - 19 | 2397 | 60 | |
XM_011543271.2 | XP_011541573.1 | 18 - 19 | 2437 | 60 | |
XM_011543272.1 | XP_011541574.1 | 18 - 19 | 2278 | 60 | |
XM_011543276.2 | XP_011541578.1 | 10 - 11 | 1400 | 60 | |
XM_017009229.1 | XP_016864718.1 | 20 - 21 | 2629 | 60 | |
XM_017009230.1 | XP_016864719.1 | 17 - 18 | 2352 | 60 | |
XM_017009231.1 | XP_016864720.1 | 17 - 18 | 4175 | 60 | |
XM_017009232.1 | XP_016864721.1 | 18 - 19 | 2270 | 60 | |
XM_017009233.1 | XP_016864722.1 | 17 - 18 | 2213 | 60 | |
GenBank mRNA | AK315234.1 | - | 16 - 17 | 2272 | 60 |
J03358.1 | - | 18 - 19 | 2584 | 60 | |
JQ412173.1 | - | 9 - 10 | 1148 | 60 |