The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014415.3 | NP_055230.2 | 1 - 2 | 541 | 85 |
GenBank mRNA | AK312859.1 | - | 1 - 2 | 533 | 85 |
BC111700.1 | - | 1 - 2 | 448 | 85 | |
U69274.1 | - | 1 - 2 | 472 | 85 |