The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006723785.2 | XP_006723848.1 | 2 - 3 | 592 | 65 |
XM_011528801.1 | XP_011527103.1 | 2 - 3 | 376 | 65 | |
XM_011528802.1 | XP_011527104.1 | 2 - 3 | 371 | 65 | |
XM_011528803.2 | XP_011527105.1 | 2 - 3 | 298 | 65 |