The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204747.1 | NP_001191676.1 | 22 - 23 | 3089 | 112 |
NM_002913.4 | NP_002904.3 | 22 - 23 | 3086 | 112 | |
XM_011513730.1 | XP_011512032.1 | 21 - 22 | 3011 | 112 | |
XM_011513731.1 | XP_011512033.1 | 20 - 21 | 2579 | 112 | |
XM_017008528.1 | XP_016864017.1 | 21 - 22 | 3008 | 112 | |
GenBank mRNA | AF040250.1 | - | 22 - 23 | 2991 | 112 |
AK291612.1 | - | 22 - 23 | 3070 | 112 | |
BC035297.1 | - | 22 - 23 | 3040 | 112 | |
BC051751.1 | - | 22 - 23 | 3056 | 112 | |
BC051786.2 | - | 22 - 23 | 3070 | 112 | |
L23320.1 | - | 22 - 23 | 3155 | 112 |