Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC2A10
Entrez Gene ID
81031
Gene Name
solute carrier family 2 member 10
Gene Aliases
ATS, GLUT10
Chromosome Location
Chr.20: 46708358 - 46736347 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030777.3 NP_110404.1 2 - 3 1382 62
XM_011529060.2 XP_011527362.1 2 - 3 1465 62
XM_011529061.2 XP_011527363.1 2 - 3 3022 62
XM_011529063.2 XP_011527365.1 2 - 3 1475 62
XM_011529065.2 XP_011527367.1 2 - 3 1472 62
XM_017028087.1 XP_016883576.1 2 - 3 1375 62
GenBank mRNA AF248053.1 - 2 - 3 1535 62
AF321240.1 - 2 - 3 1337 62
AF479407.1 - 1 - 2 36 62
AK055548.1 - 2 - 3 1382 62
AK290961.1 - 2 - 3 1373 62
AK315269.1 - 2 - 3 1373 62
AL137188.3 - 1 - 2 1293 62
BC101657.1 - 2 - 3 1381 62
BC113423.1 - 2 - 3 1381 62

View Full Product Details