The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177431.1 | NP_001170902.1 | 4 - 5 | 612 | 59 |
NM_021625.4 | NP_067638.3 | 4 - 5 | 803 | 59 | |
NM_147204.2 | NP_671737.1 | 3 - 4 | 745 | 59 | |
XM_005253918.1 | XP_005253975.1 | 4 - 5 | 1163 | 59 | |
XM_011538630.2 | XP_011536932.2 | 4 - 5 | 1906 | 59 | |
XM_011538632.2 | XP_011536934.2 | 4 - 5 | 1907 | 59 | |
XM_011538634.2 | XP_011536936.2 | 4 - 5 | 1908 | 59 | |
XM_011538635.2 | XP_011536937.1 | 4 - 5 | 1910 | 59 | |
XM_011538636.2 | XP_011536938.1 | 4 - 5 | 1911 | 59 | |
XM_017019774.1 | XP_016875263.1 | 4 - 5 | 1536 | 59 | |
GenBank mRNA | AB032427.1 | - | 3 - 4 | 809 | 59 |
AB073669.1 | - | 3 - 4 | 714 | 59 | |
AB100308.1 | - | 3 - 4 | 714 | 59 | |
AF258465.1 | - | 3 - 4 | 714 | 59 | |
AF279673.1 | - | 3 - 4 | 510 | 59 | |
AJ296305.1 | - | 4 - 5 | 803 | 59 | |
AL834192.1 | - | 4 - 5 | 775 | 59 | |
BC117426.1 | - | 4 - 5 | 797 | 59 | |
BC143307.1 | - | 4 - 5 | 797 | 59 | |
BC143315.1 | - | 4 - 5 | 797 | 59 | |
DQ059645.1 | - | 4 - 5 | 612 | 59 |