The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177428.1 | NP_001170899.1 | 4 - 5 | 1047 | 86 |
NM_001177431.1 | NP_001170902.1 | 6 - 7 | 1055 | 86 | |
NM_021625.4 | NP_067638.3 | 6 - 7 | 1246 | 86 | |
XM_005253918.1 | XP_005253975.1 | 6 - 7 | 1606 | 86 | |
XM_011538630.2 | XP_011536932.2 | 6 - 7 | 2349 | 86 | |
XM_011538631.2 | XP_011536933.2 | 5 - 6 | 2209 | 86 | |
XM_011538634.2 | XP_011536936.2 | 6 - 7 | 2351 | 86 | |
XM_011538635.2 | XP_011536937.1 | 6 - 7 | 2353 | 86 | |
XM_011538636.2 | XP_011536938.1 | 6 - 7 | 2354 | 86 | |
XM_017019774.1 | XP_016875263.1 | 6 - 7 | 1979 | 86 | |
GenBank mRNA | AB032427.1 | - | 5 - 6 | 1252 | 86 |
AB100308.1 | - | 5 - 6 | 1157 | 86 | |
AF258465.1 | - | 5 - 6 | 1157 | 86 | |
AF279673.1 | - | 5 - 6 | 953 | 86 | |
AJ296305.1 | - | 6 - 7 | 1246 | 86 | |
AL834192.1 | - | 6 - 7 | 1219 | 86 | |
BC117426.1 | - | 6 - 7 | 1240 | 86 | |
BC143307.1 | - | 6 - 7 | 1240 | 86 | |
BC143315.1 | - | 6 - 7 | 1240 | 86 | |
DQ059644.1 | - | 4 - 5 | 1016 | 86 | |
DQ059645.1 | - | 6 - 7 | 1055 | 86 |