Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UCMA
Entrez Gene ID
221044
Gene Name
upper zone of growth plate and cartilage matrix associated
Gene Aliases
C10orf49, GRP, GRP/UCMA
Chromosome Location
Chr.10: 13221766 - 13234328 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303118.1 NP_001290047.1 1 - 2 134 75
NM_145314.2 NP_660357.2 1 - 2 134 75
GenBank mRNA BC018068.1 - 1 - 2 137 75
JX169863.1 - 1 - 2 93 75
JX169864.1 - 1 - 2 93 75

View Full Product Details