The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127487.1 | NP_001120959.1 | 40 - 41 | 7109 | 104 |
NM_001458.4 | NP_001449.3 | 41 - 42 | 7208 | 104 | |
GenBank mRNA | AB208865.1 | - | 18 - 19 | 2850 | 104 |
AB371585.1 | - | 41 - 42 | 7228 | 104 | |
AF089841.1 | - | 41 - 42 | 7048 | 104 | |
AF146692.1 | - | 40 - 41 | 7112 | 104 | |
AJ012737.1 | - | 40 - 41 | 6918 | 104 |