The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002202.2 | NP_002193.2 | 3 - 4 | 1027 | 84 |
XM_011543380.2 | XP_011541682.1 | 2 - 3 | 2145 | 84 | |
GenBank mRNA | AK290742.1 | - | 3 - 4 | 745 | 84 |
BC031213.1 | - | 3 - 4 | 1028 | 84 | |
S70721.1 | - | 3 - 4 | 484 | 84 | |
U07559.1 | - | 3 - 4 | 718 | 84 |