Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTRA3
Entrez Gene ID
94031
Gene Name
HtrA serine peptidase 3
Gene Aliases
Prsp, Tasp
Chromosome Location
Chr.4: 8269757 - 8307111 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053044.4 NP_444272.1 6 - 7 1254 77
GenBank mRNA AY040094.2 - 6 - 7 1271 77
AY280665.1 - 6 - 7 1242 77
BC034390.1 - 6 - 7 1254 77
BC035717.1 - 6 - 7 1254 77

View Full Product Details