The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111125.2 | NP_001104595.1 | 4 - 5 | 1604 | 68 |
NM_015075.1 | NP_055890.1 | 4 - 5 | 881 | 68 | |
XM_006724579.2 | XP_006724642.1 | 5 - 6 | 1726 | 68 | |
XM_006724580.3 | XP_006724643.1 | 4 - 5 | 894 | 68 | |
XM_006724581.3 | XP_006724644.1 | 5 - 6 | 1726 | 68 | |
XM_006724582.3 | XP_006724645.1 | 5 - 6 | 1726 | 68 | |
XM_006724583.3 | XP_006724646.1 | 5 - 6 | 1726 | 68 | |
XM_006724584.2 | XP_006724647.1 | 5 - 6 | 1726 | 68 | |
XM_011530773.2 | XP_011529075.1 | 3 - 4 | 798 | 68 | |
XM_011530774.2 | XP_011529076.1 | 5 - 6 | 1726 | 68 | |
XM_011530776.1 | XP_011529078.1 | 5 - 6 | 1726 | 68 | |
XM_011530777.1 | XP_011529079.1 | 5 - 6 | 1726 | 68 | |
XM_017029359.1 | XP_016884848.1 | 3 - 4 | 1600 | 68 | |
XM_017029360.1 | XP_016884849.1 | 4 - 5 | 1012 | 68 | |
GenBank mRNA | AB011094.1 | - | 4 - 5 | 1604 | 68 |
AK095232.1 | - | 4 - 5 | 881 | 68 | |
FJ154890.1 | - | 3 - 4 | 783 | 68 | |
FJ154891.1 | - | 4 - 5 | 874 | 68 |