The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003370.3 | NP_003361.1 | 11 - 12 | 1387 | 69 |
XM_005259199.1 | XP_005259256.1 | 11 - 12 | 1372 | 69 | |
XM_005259200.1 | XP_005259257.1 | 11 - 12 | 1372 | 69 | |
XM_017027200.1 | XP_016882689.1 | 11 - 12 | 1369 | 69 | |
GenBank mRNA | AK314812.1 | - | 11 - 12 | 1354 | 69 |
BC026019.1 | - | 11 - 12 | 1324 | 69 | |
BC038224.1 | - | 11 - 12 | 1305 | 69 | |
KU178381.1 | - | 10 - 11 | 1045 | 69 | |
KU178382.1 | - | 11 - 12 | 889 | 69 | |
KU178383.1 | - | 11 - 12 | 886 | 69 | |
KU178384.1 | - | 10 - 11 | 769 | 69 | |
KU178385.1 | - | 8 - 9 | 594 | 69 | |
Z46389.1 | - | 11 - 12 | 1299 | 69 |