Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VASP
Entrez Gene ID
7408
Gene Name
vasodilator-stimulated phosphoprotein
Gene Aliases
-
Chromosome Location
Chr.19: 45507430 - 45526989 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003370.3 NP_003361.1 4 - 5 773 143
XM_005259199.1 XP_005259256.1 4 - 5 761 143
XM_005259200.1 XP_005259257.1 4 - 5 761 143
XM_017027200.1 XP_016882689.1 4 - 5 761 143
GenBank mRNA AK314812.1 - 4 - 5 740 143
BC026019.1 - 4 - 5 713 143
BC038224.1 - 4 - 5 691 143
KU178381.1 - 3 - 4 431 143
KU178382.1 - 3 - 4 431 143
KU178383.1 - 3 - 4 431 143
KU178384.1 - 3 - 4 431 143
Z46389.1 - 4 - 5 685 143

View Full Product Details