The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170634.1 | NP_001164105.1 | 1 - 2 | 117 | 76 |
NM_001170937.1 | NP_001164408.1 | 1 - 2 | 117 | 76 | |
NM_004960.3 | NP_004951.1 | 1 - 2 | 117 | 76 | |
NR_028388.2 | 1 - 2 | 117 | 76 | ||
XM_005255233.4 | XP_005255290.1 | 1 - 2 | 783 | 76 | |
XM_011545781.1 | XP_011544083.1 | 1 - 2 | 89 | 76 | |
GenBank mRNA | AB208902.1 | - | 1 - 2 | 77 | 76 |
AK098774.1 | - | 1 - 2 | 88 | 76 | |
AK290936.1 | - | 1 - 2 | 88 | 76 | |
AK299129.1 | - | 1 - 2 | 86 | 76 | |
AK301110.1 | - | 1 - 2 | 88 | 76 | |
BC000402.2 | - | 1 - 2 | 80 | 76 | |
BC002459.1 | - | 1 - 2 | 52 | 76 | |
BC026062.2 | - | 1 - 2 | 70 | 76 | |
BX423519.2 | - | 1 - 2 | 96 | 77 | |
DA541747.1 | - | 1 - 2 | 88 | 76 | |
DC361263.1 | - | 1 - 2 | 117 | 76 | |
S62140.1 | - | 1 - 2 | 90 | 76 | |
U00942.1 | - | - | 50 | 74 | |
X71428.1 | - | 1 - 2 | 84 | 76 |