Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAT8
Entrez Gene ID
84148
Gene Name
lysine acetyltransferase 8
Gene Aliases
MOF, MYST1, ZC2HC8, hMOF
Chromosome Location
Chr.16: 31117664 - 31131393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032188.2 NP_115564.2 3 - 4 479 91
NM_182958.2 NP_892003.2 3 - 4 479 91
XM_011545970.1 XP_011544272.1 4 - 5 1020 91
XM_011545971.1 XP_011544273.1 4 - 5 1025 91
GenBank mRNA AF217501.1 - 3 - 4 445 91
AF260665.2 - 3 - 4 377 91
AK021872.1 - 3 - 4 476 91
AK024102.1 - 3 - 4 477 91
AK225660.1 - 3 - 4 479 91
AK291106.1 - 3 - 4 476 91
BC037773.1 - 3 - 4 476 91
BP342476.1 - 3 - 4 479 91

View Full Product Details