The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000932.2 | NP_000923.1 | 10 - 11 | 1139 | 83 |
NM_001184883.1 | NP_001171812.1 | 8 - 9 | 938 | 83 | |
NM_001316314.1 | NP_001303243.1 | 10 - 11 | 1139 | 83 | |
XM_011545101.2 | XP_011543403.1 | 10 - 11 | 1127 | 83 | |
XM_017017925.1 | XP_016873414.1 | 10 - 11 | 1127 | 83 | |
GenBank mRNA | BC032659.1 | - | 8 - 9 | 885 | 83 |
BC142681.1 | - | 10 - 11 | 1092 | 83 | |
BC146645.1 | - | 10 - 11 | 1115 | 83 | |
Z16411.1 | - | 4 - 5 | 464 | 83 | |
Z26649.1 | - | 10 - 11 | 1142 | 83 |