The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000932.2 | NP_000923.1 | 12 - 13 | 1466 | 80 |
NM_001184883.1 | NP_001171812.1 | 10 - 11 | 1265 | 80 | |
NM_001316314.1 | NP_001303243.1 | 12 - 13 | 1466 | 80 | |
XM_011545101.2 | XP_011543403.1 | 12 - 13 | 1454 | 80 | |
XM_017017925.1 | XP_016873414.1 | 12 - 13 | 1454 | 80 | |
GenBank mRNA | BC032659.1 | - | 10 - 11 | 1212 | 80 |
BC142681.1 | - | 12 - 13 | 1419 | 80 | |
BC146645.1 | - | 12 - 13 | 1442 | 80 | |
Z16411.1 | - | 6 - 7 | 791 | 80 | |
Z26649.1 | - | 12 - 13 | 1469 | 80 |