The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000932.2 | NP_000923.1 | 24 - 25 | 2970 | 74 |
NM_001184883.1 | NP_001171812.1 | 22 - 23 | 2769 | 74 | |
NM_001316314.1 | NP_001303243.1 | 24 - 25 | 2970 | 74 | |
XM_011545101.2 | XP_011543403.1 | 24 - 25 | 2958 | 74 | |
GenBank mRNA | BC032659.1 | - | 22 - 23 | 2716 | 74 |
BC041625.1 | - | 6 - 7 | 633 | 74 | |
BC142681.1 | - | 24 - 25 | 2923 | 74 | |
BC146645.1 | - | 24 - 25 | 2946 | 74 | |
Z16411.1 | - | 18 - 19 | 2295 | 74 | |
Z26649.1 | - | 24 - 25 | 2973 | 74 |