The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206739.1 | NP_001193668.1 | 9 - 10 | 785 | 101 |
NM_014288.4 | NP_055103.3 | 8 - 9 | 668 | 101 | |
XM_017000855.1 | XP_016856344.1 | 7 - 8 | 681 | 101 | |
GenBank mRNA | AA873717.1 | - | 1 - 2 | 101 | |
AF175306.1 | - | 8 - 9 | 559 | 101 | |
BC009929.2 | - | 8 - 9 | 575 | 101 | |
BC014385.1 | - | 9 - 10 | 710 | 101 |