The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020196.2 | NP_064581.2 | 18 - 19 | 2539 | 87 |
GenBank mRNA | AB026111.1 | - | 18 - 19 | 2528 | 87 |
AB033003.1 | - | 16 - 17 | 2215 | 87 | |
AF226051.1 | - | 18 - 19 | 2539 | 87 | |
AF258567.1 | - | 18 - 19 | 2534 | 87 | |
AF272147.1 | - | 18 - 19 | 2549 | 87 | |
AK025858.1 | - | 13 - 14 | 1822 | 87 | |
BC007208.1 | - | 18 - 19 | 2521 | 87 | |
BC008778.1 | - | 13 - 14 | 1854 | 87 | |
CR749864.1 | - | 15 - 16 | 2068 | 87 |