The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020196.2 | NP_064581.2 | 8 - 9 | 1144 | 137 |
GenBank mRNA | AB026111.1 | - | 8 - 9 | 1133 | 137 |
AB033003.1 | - | 6 - 7 | 820 | 137 | |
AF226051.1 | - | 8 - 9 | 1143 | 137 | |
AF258567.1 | - | 8 - 9 | 1139 | 137 | |
AF272147.1 | - | 8 - 9 | 1154 | 137 | |
AK025858.1 | - | 3 - 4 | 427 | 137 | |
AK074035.1 | - | 4 - 5 | 1715 | 137 | |
BC007208.1 | - | 8 - 9 | 1126 | 137 | |
BC008778.1 | - | 3 - 4 | 459 | 137 | |
CR749864.1 | - | 5 - 6 | 673 | 137 |