Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPP2
Entrez Gene ID
6694
Gene Name
secreted phosphoprotein 2
Gene Aliases
SPP-24, SPP24
Chromosome Location
Chr.2: 234050671 - 234077134 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006944.2 NP_008875.1 3 - 4 422 63
XM_005246102.3 XP_005246159.1 3 - 4 453 63
XM_011511698.2 XP_011510000.1 3 - 4 453 63
XM_011511699.2 XP_011510001.1 3 - 4 453 63
XM_011511700.2 XP_011510002.1 3 - 4 453 63
GenBank mRNA AJ308099.1 - 3 - 4 422 63
BC069401.1 - 3 - 4 337 63
BC106705.1 - 3 - 4 337 63
BC112438.1 - 3 - 4 337 63
BC113972.1 - 2 - 3 212 63
U20530.1 - 3 - 4 337 63

View Full Product Details