Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALKBH7
Entrez Gene ID
84266
Gene Name
alkB homolog 7
Gene Aliases
ABH7, SPATA11, UNQ6002
Chromosome Location
Chr.19: 6372433 - 6375250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032306.3 NP_115682.1 2 - 3 771 84
XM_005259658.4 XP_005259715.1 2 - 3 347 84
XM_017027355.1 XP_016882844.1 2 - 3 407 84
GenBank mRNA AK223336.1 - 2 - 3 394 84
AK311956.1 - 2 - 3 410 84
AY358858.1 - 2 - 3 771 84
AY427650.2 - 2 - 3 771 84
BC004393.1 - 2 - 3 426 84

View Full Product Details