Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPB7
Entrez Gene ID
27129
Gene Name
heat shock protein family B (small) member 7
Gene Aliases
cvHSP
Chromosome Location
Chr.1: 16014028 - 16018796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014424.4 NP_055239.1 1 - 2 1024 97
GenBank mRNA AF155908.1 - 1 - 2 271 97
AJ243191.1 - 1 - 2 271 97
AK057295.1 - 1 - 2 1022 97
AK092275.1 - 2 - 3 547 97
AK095911.1 - 1 - 2 277 97
BC006319.2 - 1 - 2 690 97
BX537990.1 - 1 - 2 276 97

View Full Product Details