The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193509.1 | NP_001180438.1 | 10 - 11 | 1258 | 80 |
NM_017735.4 | NP_060205.3 | 10 - 11 | 1505 | 80 | |
XM_005264416.1 | XP_005264473.1 | 10 - 11 | 1492 | 80 | |
XM_011532958.1 | XP_011531260.1 | 10 - 11 | 1492 | 80 | |
GenBank mRNA | AK000279.1 | - | 7 - 8 | 713 | 80 |
AK000665.1 | - | 5 - 6 | 497 | 80 | |
AK027570.1 | - | 10 - 11 | 1414 | 80 | |
AK225295.1 | - | 7 - 8 | 713 | 80 | |
AK299196.1 | - | 10 - 11 | 1247 | 80 | |
BC001248.1 | - | 9 - 10 | 1038 | 80 | |
BC063791.1 | - | 10 - 11 | 1463 | 80 |