The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193509.1 | NP_001180438.1 | 2 - 3 | 290 | 95 |
NM_017735.4 | NP_060205.3 | 2 - 3 | 537 | 95 | |
XM_005264416.1 | XP_005264473.1 | 2 - 3 | 524 | 95 | |
XM_011532958.1 | XP_011531260.1 | 2 - 3 | 524 | 95 | |
GenBank mRNA | AK027570.1 | - | 2 - 3 | 446 | 95 |
AK299196.1 | - | 2 - 3 | 279 | 95 | |
BC001248.1 | - | 1 - 2 | 70 | 95 | |
BC063791.1 | - | 2 - 3 | 495 | 95 |