The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193509.1 | NP_001180438.1 | 5 - 6 | 667 | 99 |
NM_017735.4 | NP_060205.3 | 5 - 6 | 914 | 99 | |
XM_005264416.1 | XP_005264473.1 | 5 - 6 | 901 | 99 | |
XM_011532958.1 | XP_011531260.1 | 5 - 6 | 901 | 99 | |
GenBank mRNA | AK000279.1 | - | 2 - 3 | 122 | 99 |
AK027570.1 | - | 5 - 6 | 823 | 99 | |
AK225295.1 | - | 2 - 3 | 122 | 99 | |
AK299196.1 | - | 5 - 6 | 656 | 99 | |
BC001248.1 | - | 4 - 5 | 447 | 99 | |
BC063791.1 | - | 5 - 6 | 872 | 99 |