Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BIRC6
Entrez Gene ID
57448
Gene Name
baculoviral IAP repeat containing 6
Gene Aliases
APOLLON, BRUCE
Chromosome Location
Chr.2: 32356907 - 32618899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016252.3 NP_057336.3 42 - 43 8227 108
XM_005264449.4 XP_005264506.2 42 - 43 8429 108
XM_005264450.4 XP_005264507.2 42 - 43 8417 108
XM_005264451.4 XP_005264508.2 42 - 43 8402 108
XM_005264452.4 XP_005264509.2 42 - 43 8132 108
XM_005264453.4 XP_005264510.2 42 - 43 8105 108
XM_005264454.4 XP_005264511.2 42 - 43 8429 108
XM_005264455.4 XP_005264512.2 41 - 42 8297 108
XM_006712054.3 XP_006712117.1 42 - 43 8411 108
XM_006712055.3 XP_006712118.1 42 - 43 8429 108
XM_006712056.3 XP_006712119.1 42 - 43 8330 108
XM_011533003.2 XP_011531305.1 42 - 43 7874 108
XM_011533005.2 XP_011531307.1 42 - 43 8429 108
XM_017004556.1 XP_016860045.1 42 - 43 8411 108
XM_017004557.1 XP_016860046.1 42 - 43 8318 108
XM_017004558.1 XP_016860047.1 42 - 43 8288 108
XM_017004559.1 XP_016860048.1 42 - 43 8429 108
XM_017004560.1 XP_016860049.1 42 - 43 8411 108
GenBank mRNA AB033115.2 - 13 - 14 1907 108
AF265555.1 - 42 - 43 8009 108

View Full Product Details