Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BIRC6
Entrez Gene ID
57448
Gene Name
baculoviral IAP repeat containing 6
Gene Aliases
APOLLON, BRUCE
Chromosome Location
Chr.2: 32356907 - 32618899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016252.3 NP_057336.3 49 - 50 9635 78
XM_005264449.4 XP_005264506.2 49 - 50 9837 78
XM_005264450.4 XP_005264507.2 49 - 50 9825 78
XM_005264451.4 XP_005264508.2 49 - 50 9810 78
XM_005264452.4 XP_005264509.2 49 - 50 9540 78
XM_005264453.4 XP_005264510.2 49 - 50 9513 78
XM_005264454.4 XP_005264511.2 49 - 50 9837 78
XM_005264455.4 XP_005264512.2 48 - 49 9705 78
XM_006712054.3 XP_006712117.1 49 - 50 9819 78
XM_006712055.3 XP_006712118.1 49 - 50 9837 78
XM_006712056.3 XP_006712119.1 49 - 50 9738 78
XM_011533003.2 XP_011531305.1 49 - 50 9282 78
XM_011533005.2 XP_011531307.1 49 - 50 9837 78
XM_017004556.1 XP_016860045.1 49 - 50 9819 78
XM_017004557.1 XP_016860046.1 49 - 50 9726 78
XM_017004558.1 XP_016860047.1 49 - 50 9696 78
XM_017004559.1 XP_016860048.1 49 - 50 9837 78
XM_017004560.1 XP_016860049.1 49 - 50 9819 78
GenBank mRNA AB033115.2 - 20 - 21 3315 78
AF265555.1 - 49 - 50 9417 78
AK023848.1 - 4 - 5 587 78

View Full Product Details