Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BIRC6
Entrez Gene ID
57448
Gene Name
baculoviral IAP repeat containing 6
Gene Aliases
APOLLON, BRUCE
Chromosome Location
Chr.2: 32356907 - 32618899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016252.3 NP_057336.3 51 - 52 10118 139
XM_005264449.4 XP_005264506.2 51 - 52 10320 139
XM_005264450.4 XP_005264507.2 51 - 52 10308 139
XM_005264451.4 XP_005264508.2 51 - 52 10293 139
XM_005264452.4 XP_005264509.2 51 - 52 10023 139
XM_005264453.4 XP_005264510.2 51 - 52 9996 139
XM_005264454.4 XP_005264511.2 51 - 52 10320 139
XM_005264455.4 XP_005264512.2 50 - 51 10188 139
XM_006712054.3 XP_006712117.1 51 - 52 10302 139
XM_006712055.3 XP_006712118.1 51 - 52 10320 139
XM_006712056.3 XP_006712119.1 51 - 52 10221 139
XM_011533003.2 XP_011531305.1 51 - 52 9765 139
XM_011533005.2 XP_011531307.1 51 - 52 10320 139
XM_017004556.1 XP_016860045.1 51 - 52 10302 139
XM_017004557.1 XP_016860046.1 51 - 52 10209 139
XM_017004558.1 XP_016860047.1 51 - 52 10179 139
XM_017004559.1 XP_016860048.1 51 - 52 10320 139
XM_017004560.1 XP_016860049.1 51 - 52 10302 139
GenBank mRNA AB033115.2 - 22 - 23 3798 139
AF265555.1 - 51 - 52 9900 139
AK023848.1 - 6 - 7 1070 139

View Full Product Details