The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001973.2 | NP_001001973.1 | 5 - 6 | 676 | 138 |
NM_001320886.1 | NP_001307815.1 | 4 - 5 | 641 | 138 | |
NM_005174.3 | NP_005165.1 | 5 - 6 | 676 | 138 | |
XM_017016290.1 | XP_016871779.1 | 4 - 5 | 586 | 138 | |
GenBank mRNA | AK292896.1 | - | 5 - 6 | 603 | 138 |
AK296803.1 | - | 4 - 5 | 568 | 138 | |
BC000470.2 | - | 5 - 6 | 591 | 138 | |
BC000931.3 | - | 5 - 6 | 624 | 138 | |
BC013394.2 | - | 5 - 6 | 591 | 138 | |
BC013865.1 | - | 5 - 6 | 589 | 138 | |
BC016812.1 | - | 5 - 6 | 606 | 138 | |
BC020824.1 | - | 5 - 6 | 612 | 138 | |
BC026049.1 | - | 5 - 6 | 551 | 138 | |
BI603806.1 | - | 4 - 5 | 606 | 138 | |
BQ893894.1 | - | 5 - 6 | 651 | 138 | |
CR457403.1 | - | 5 - 6 | 572 | 138 | |
D16562.1 | - | 5 - 6 | 603 | 138 | |
D16563.1 | - | 5 - 6 | 603 | 138 |