The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256160.1 | NP_001243089.1 | 6 - 7 | 1365 | 147 |
NM_024997.3 | NP_079273.2 | 5 - 6 | 1322 | 147 | |
XM_006720953.3 | XP_006721016.1 | 4 - 5 | 1149 | 147 | |
XM_017023705.1 | XP_016879194.1 | 4 - 5 | 2290 | 147 | |
XM_017023706.1 | XP_016879195.1 | 4 - 5 | 2290 | 147 | |
XM_017023707.1 | XP_016879196.1 | 4 - 5 | 2290 | 147 | |
XM_017023708.1 | XP_016879197.1 | 3 - 4 | 373 | 147 | |
GenBank mRNA | AK022730.1 | - | 1 - 2 | 82 | 147 |
AK093414.1 | - | 6 - 7 | 1363 | 147 | |
AK223492.1 | - | 1 - 2 | 133 | 147 | |
AY560615.1 | - | 3 - 4 | 1092 | 147 | |
BC033891.1 | - | 5 - 6 | 1318 | 147 | |
BC069695.1 | - | 1 - 2 | 59 | 147 | |
BC069713.1 | - | 1 - 2 | 59 | 147 | |
BC069730.1 | - | 1 - 2 | 59 | 147 | |
BC137079.1 | - | 4 - 5 | 1221 | 147 |