The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143685.1 | NP_001137157.1 | 2 - 3 | 432 | 95 |
NM_001190158.1 | NP_001177087.1 | 3 - 4 | 516 | 95 | |
NM_145024.2 | NP_659461.1 | 2 - 3 | 432 | 95 | |
GenBank mRNA | AK056109.1 | - | 2 - 3 | 432 | 95 |
AK294334.1 | - | 3 - 4 | 516 | 95 | |
AY907669.1 | - | 2 - 3 | 359 | 95 | |
BC039073.1 | - | 2 - 3 | 313 | 95 | |
BC069501.1 | - | 2 - 3 | 403 | 95 | |
BC069548.1 | - | 2 - 3 | 402 | 95 | |
BC117126.1 | - | 2 - 3 | 405 | 95 | |
BC143692.1 | - | 2 - 3 | 405 | 95 | |
EU794678.1 | - | 2 - 3 | 432 | 95 |