The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271015.1 | NP_001257944.1 | 1 - 2 | 103 | 85 |
NM_018299.4 | NP_060769.4 | 1 - 2 | 103 | 85 | |
NR_073119.1 | 1 - 2 | 103 | 85 | ||
NR_073120.1 | 1 - 2 | 103 | 85 | ||
NR_073121.1 | 1 - 2 | 103 | 85 | ||
GenBank mRNA | AK295792.1 | - | 1 - 2 | 103 | 85 |
BC016326.1 | - | 1 - 2 | 57 | 85 | |
BU155172.1 | - | 1 - 2 | 49 | 85 | |
DA352184.1 | - | 1 - 2 | 55 | 85 |