The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012082.3 | NP_036214.2 | 6 - 7 | 759 | 65 |
XM_011516947.2 | XP_011515249.1 | 6 - 7 | 4358 | 65 | |
XM_011516948.2 | XP_011515250.1 | 5 - 6 | 660 | 65 | |
GenBank mRNA | AF119334.1 | - | 6 - 7 | 760 | 65 |
AK296997.1 | - | 6 - 7 | 908 | 65 | |
BC109222.1 | - | 6 - 7 | 760 | 65 | |
BC109223.1 | - | 6 - 7 | 760 | 65 |