Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSPAN8
Entrez Gene ID
7103
Gene Name
tetraspanin 8
Gene Aliases
CO-029, TM4SF3
Chromosome Location
Chr.12: 71125093 - 71157999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004616.2 NP_004607.1 5 - 6 522 90
XM_006719583.3 XP_006719646.1 5 - 6 476 90
XM_017019913.1 XP_016875402.1 4 - 5 558 90
GenBank mRNA AK223022.1 - 4 - 5 450 90
AK313112.1 - 4 - 5 505 90
BC005246.1 - 5 - 6 522 90
BC070168.1 - 9 - 10 896 90
M35252.1 - 4 - 5 480 90

View Full Product Details