Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHCBP1L
Entrez Gene ID
81626
Gene Name
SHC binding and spindle associated 1 like
Gene Aliases
C1orf14, GE36
Chromosome Location
Chr.1: 182899865 - 182953418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030933.2 NP_112195.2 9 - 10 1993 113
GenBank mRNA AF288397.1 - 10 - 11 1651 113
AF288398.1 - 9 - 10 1992 113
BC026084.1 - 8 - 9 2173 113
BC050305.1 - 9 - 10 1945 113
BC132764.1 - 9 - 10 1969 113
BC144326.1 - 9 - 10 1965 113

View Full Product Details