The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101653.1 | NP_001095123.1 | 2 - 3 | 232 | 85 |
NM_001307937.1 | NP_001294866.1 | 4 - 5 | 383 | 85 | |
NM_014186.3 | NP_054905.2 | 3 - 4 | 358 | 85 | |
GenBank mRNA | AF161515.1 | - | 3 - 4 | 339 | 85 |
AK055668.1 | - | 3 - 4 | 330 | 85 | |
AK223213.1 | - | 3 - 4 | 337 | 85 | |
AK290065.1 | - | 3 - 4 | 85 | ||
AK298978.1 | - | 3 - 4 | 337 | 85 | |
AK314477.1 | - | 3 - 4 | 327 | 85 | |
AL136688.1 | - | 3 - 4 | 332 | 85 | |
AY542164.1 | - | 3 - 4 | 321 | 85 | |
BC010892.1 | - | 3 - 4 | 337 | 85 | |
BG702189.1 | - | 2 - 3 | 237 | 85 | |
BM172201.1 | - | 2 - 3 | 237 | 85 |