Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSD4
Entrez Gene ID
23550
Gene Name
pleckstrin and Sec7 domain containing 4
Gene Aliases
EFA6B, TIC
Chromosome Location
Chr.2: 113157399 - 113203229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012455.2 NP_036587.2 10 - 11 2365 68
XM_005263634.2 XP_005263691.1 10 - 11 2430 68
XM_006712392.2 XP_006712455.1 10 - 11 2377 68
XM_006712393.2 XP_006712456.1 10 - 11 2377 68
XM_006712394.2 XP_006712457.1 10 - 11 2377 68
GenBank mRNA AB208798.1 - 9 - 10 2804 68
AJ459781.1 - 9 - 10 2203 68
AK023421.1 - 8 - 9 1254 68
AK290045.1 - 10 - 11 2375 68
BC035307.1 - 10 - 11 2267 68
BC073151.1 - 10 - 11 2327 68
U63127.1 - 9 - 10 2245 68

View Full Product Details