The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012455.2 | NP_036587.2 | 15 - 16 | 2949 | 70 |
XM_005263634.2 | XP_005263691.1 | 15 - 16 | 3014 | 70 | |
XM_006712392.2 | XP_006712455.1 | 15 - 16 | 2958 | 70 | |
GenBank mRNA | AB208798.1 | - | 14 - 15 | 3385 | 70 |
AJ459781.1 | - | 14 - 15 | 2787 | 70 | |
AK023421.1 | - | 13 - 14 | 1838 | 70 | |
AK094598.1 | - | 2 - 3 | 861 | 70 | |
AK290045.1 | - | 15 - 16 | 2959 | 70 | |
AK294268.1 | - | 3 - 4 | 599 | 70 | |
BC035307.1 | - | 15 - 16 | 2848 | 70 | |
BC073151.1 | - | 15 - 16 | 2911 | 70 | |
U63127.1 | - | 14 - 15 | 2829 | 70 |