Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7orf57
Entrez Gene ID
136288
Gene Name
chromosome 7 open reading frame 57
Gene Aliases
-
Chromosome Location
Chr.7: 48035511 - 48061297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100159.2 NP_001093629.1 5 - 6 724 78
NM_001267865.1 NP_001254794.1 4 - 5 568 78
NM_001267866.1 NP_001254795.1 4 - 5 568 78
XM_006715649.3 XP_006715712.1 4 - 5 584 78
XM_006715650.2 XP_006715713.1 4 - 5 551 78
XM_011515121.1 XP_011513423.1 5 - 6 628 78
XM_011515122.2 XP_011513424.1 4 - 5 584 78
XM_011515123.2 XP_011513425.1 4 - 5 584 78
GenBank mRNA BC031107.1 - 5 - 6 715 78
BC045737.1 - 4 - 5 568 78
BI560654.1 - 4 - 5 564 77

View Full Product Details