The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015030.1 | NP_055845.1 | 15 - 16 | 1855 | 171 |
XM_005248090.4 | XP_005248147.1 | 16 - 17 | 1913 | 171 | |
XM_011513680.2 | XP_011511982.1 | 13 - 14 | 1375 | 171 | |
XM_011513681.2 | XP_011511983.1 | 13 - 14 | 1375 | 171 | |
XM_011513682.2 | XP_011511984.1 | 12 - 13 | 1330 | 171 | |
XM_011513683.2 | XP_011511985.1 | 15 - 16 | 1749 | 171 | |
XM_011513685.2 | XP_011511987.1 | 13 - 14 | 1375 | 171 | |
XM_017008046.1 | XP_016863535.1 | 16 - 17 | 2180 | 171 |