The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015030.1 | NP_055845.1 | 32 - 33 | 4295 | 145 |
XM_005248090.4 | XP_005248147.1 | 33 - 34 | 4353 | 145 | |
XM_005248093.4 | XP_005248150.1 | 7 - 8 | 980 | 145 | |
XM_011513680.2 | XP_011511982.1 | 30 - 31 | 3815 | 145 | |
XM_011513681.2 | XP_011511983.1 | 30 - 31 | 3815 | 145 | |
XM_011513682.2 | XP_011511984.1 | 29 - 30 | 3770 | 145 | |
XM_011513683.2 | XP_011511985.1 | 32 - 33 | 4189 | 145 | |
XM_011513684.2 | XP_011511986.1 | 7 - 8 | 986 | 145 | |
XM_011513685.2 | XP_011511987.1 | 30 - 31 | 3815 | 145 | |
XM_017008046.1 | XP_016863535.1 | 33 - 34 | 4620 | 145 | |
GenBank mRNA | CR627228.1 | - | 16 - 17 | 2341 | 145 |