The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015030.1 | NP_055845.1 | 33 - 34 | 4466 | 67 |
XM_005248090.4 | XP_005248147.1 | 34 - 35 | 4524 | 67 | |
XM_005248093.4 | XP_005248150.1 | 8 - 9 | 1151 | 67 | |
XM_011513680.2 | XP_011511982.1 | 31 - 32 | 3986 | 67 | |
XM_011513681.2 | XP_011511983.1 | 31 - 32 | 3986 | 67 | |
XM_011513682.2 | XP_011511984.1 | 30 - 31 | 3941 | 67 | |
XM_011513683.2 | XP_011511985.1 | 33 - 34 | 4360 | 67 | |
XM_011513684.2 | XP_011511986.1 | 8 - 9 | 1157 | 67 | |
XM_011513685.2 | XP_011511987.1 | 31 - 32 | 3986 | 67 | |
XM_017008046.1 | XP_016863535.1 | 34 - 35 | 4791 | 67 | |
GenBank mRNA | CR627228.1 | - | 17 - 18 | 2512 | 67 |