The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318916.1 | NP_001305845.1 | 2 - 3 | 365 | 142 |
NM_018453.3 | NP_060923.2 | 2 - 3 | 365 | 142 | |
XM_011536958.1 | XP_011535260.1 | 2 - 3 | 643 | 142 | |
XM_017021460.1 | XP_016876949.1 | 2 - 3 | 365 | 142 | |
GenBank mRNA | AF217512.1 | - | 2 - 3 | 351 | 142 |
AK225467.1 | - | 2 - 3 | 320 | 142 | |
AK225477.1 | - | 2 - 3 | 329 | 142 | |
AY157300.1 | - | 2 - 3 | 347 | 142 | |
AY157301.1 | - | 2 - 3 | 347 | 142 | |
AY869694.1 | - | 2 - 3 | 288 | 142 | |
BC001245.1 | - | 2 - 3 | 338 | 142 | |
BU662131.1 | - | 2 - 3 | 313 | 142 | |
CD702290.1 | - | 2 - 3 | 394 | 142 |