Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EAPP
Entrez Gene ID
55837
Gene Name
E2F associated phosphoprotein
Gene Aliases
BM036, C14orf11
Chromosome Location
Chr.14: 34515929 - 34539737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018453.3 NP_060923.2 3 - 4 455 118
XM_011536958.1 XP_011535260.1 3 - 4 733 118
XM_017021460.1 XP_016876949.1 3 - 4 455 118
GenBank mRNA AF217512.1 - 3 - 4 441 118
AK000585.1 - 3 - 4 419 118
AK225467.1 - 3 - 4 410 118
AK225477.1 - 3 - 4 419 118
AK225991.1 - 3 - 4 419 118
AY157300.1 - 3 - 4 437 118
AY157301.1 - 3 - 4 437 118
AY869694.1 - 3 - 4 378 118
BC001245.1 - 3 - 4 428 118
CD702290.1 - 3 - 4 484 118

View Full Product Details