Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP2
Entrez Gene ID
4133
Gene Name
microtubule associated protein 2
Gene Aliases
MAP2A, MAP2B, MAP2C
Chromosome Location
Chr.2: 209424047 - 209734118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039538.1 NP_001034627.1 10 - 11 1288 102
NM_002374.3 NP_002365.3 11 - 12 4982 102
NM_031845.2 NP_114033.2 8 - 9 914 102
NM_031847.2 NP_114035.2 8 - 9 914 102
XM_005246555.1 XP_005246612.1 13 - 14 5518 102
XM_005246565.1 XP_005246622.1 9 - 10 4807 102
XM_005246566.1 XP_005246623.1 9 - 10 4795 102
XM_006712532.1 XP_006712595.1 13 - 14 5515 102
XM_011511186.1 XP_011509488.1 13 - 14 5425 102
XM_011511187.1 XP_011509489.1 12 - 13 5319 102
XM_011511188.1 XP_011509490.1 12 - 13 5319 102
XM_011511190.1 XP_011509492.1 12 - 13 5322 102
XM_011511191.1 XP_011509493.1 11 - 12 5187 102
XM_011511192.1 XP_011509494.1 11 - 12 5151 102
XM_011511193.1 XP_011509495.1 11 - 12 5148 102
XM_011511195.1 XP_011509497.1 11 - 12 5073 102
XM_011511196.2 XP_011509498.1 12 - 13 5260 102
XM_011511197.2 XP_011509499.1 12 - 13 5257 102
XM_017004111.1 XP_016859600.1 11 - 12 5148 102
XM_017004112.1 XP_016859601.1 12 - 13 5272 102
XM_017004113.1 XP_016859602.1 12 - 13 5164 102
XM_017004114.1 XP_016859603.1 11 - 12 5101 102
XM_017004115.1 XP_016859604.1 10 - 11 4893 102
XM_017004116.1 XP_016859605.1 10 - 11 4890 102
XM_017004117.1 XP_016859606.1 10 - 11 4890 102
XM_017004118.1 XP_016859607.1 11 - 12 5008 102
XM_017004119.1 XP_016859608.1 11 - 12 5098 102
XM_017004120.1 XP_016859609.1 11 - 12 5098 102
XM_017004121.1 XP_016859610.1 11 - 12 5005 102
XM_017004122.1 XP_016859611.1 11 - 12 5005 102
XM_017004123.1 XP_016859612.1 11 - 12 5089 102
XM_017004124.1 XP_016859613.1 11 - 12 4996 102
XM_017004125.1 XP_016859614.1 11 - 12 5086 102
XM_017004126.1 XP_016859615.1 11 - 12 5086 102
XM_017004127.1 XP_016859616.1 11 - 12 5086 102
XM_017004128.1 XP_016859617.1 10 - 11 4887 102
XM_017004129.1 XP_016859618.1 10 - 11 4884 102
XM_017004130.1 XP_016859619.1 9 - 10 5244 102
XM_017004131.1 XP_016859620.1 8 - 9 4636 102
XM_017004132.1 XP_016859621.1 8 - 9 4624 102
XM_017004133.1 XP_016859622.1 8 - 9 4636 102
XM_017004134.1 XP_016859623.1 8 - 9 4624 102
XM_017004135.1 XP_016859624.1 8 - 9 5070 102
XM_017004136.1 XP_016859625.1 8 - 9 4633 102
XM_017004137.1 XP_016859626.1 8 - 9 4621 102
XM_017004138.1 XP_016859627.1 9 - 10 1111 102
XM_017004139.1 XP_016859628.1 6 - 7 868 102
XM_017004140.1 XP_016859629.1 8 - 9 940 102
GenBank mRNA AB209330.1 - 7 - 8 832 102
AK291446.1 - 5 - 6 703 102
BC027583.1 - 8 - 9 986 102
BC038857.1 - 10 - 11 1277 102
BC110423.1 - 8 - 9 1162 102
BC117123.1 - 6 - 7 743 102
BC143245.1 - 6 - 7 748 102
U89329.1 - 5 - 6 666 102
U89330.1 - 8 - 9 4722 102

View Full Product Details